Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203141.1 | NP_001190070.1 | 2 - 3 | 261 | 105 | |
NM_115080.3 | NP_190788.1 | 2 - 3 | 338 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |