Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115089.3 | NP_190797.1 | 4 - 5 | 767 | 67 | |
GenBank mRNA | BT002936.1 | - | 4 - 5 | 716 | 67 | |
BT005639.1 | - | 2 - 3 | 368 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |