Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115141.3 | NP_190849.1 | 2 - 3 | 547 | 70 | |
GenBank mRNA | AF492667.1 | - | 2 - 3 | 454 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |