Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115145.2 | NP_190853.1 | 6 - 7 | 1544 | 69 | |
GenBank mRNA | U86700.1 | - | 6 - 7 | 1463 | 69 | |
Y07917.1 | - | 1473 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |