Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115151.3 | NP_190859.2 | 1 - 2 | 466 | 124 | |
GenBank mRNA | AY102637.1 | - | 1 - 2 | 246 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |