Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115180.3 | NP_566980.1 | 1 - 2 | 233 | 67 | |
GenBank mRNA | AK117256.1 | - | 1 - 2 | 188 | 67 | |
AY519599.1 | - | 1 - 2 | 120 | 67 | ||
BT005257.1 | - | 1 - 2 | 120 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |