Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115208.4 | NP_190916.1 | 12 - 13 | 2227 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |