Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHX20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339628.1 | NP_001325440.1 | 1 - 2 | 1539 | 99 | |
NM_115232.3 | NP_190940.1 | 2 - 3 | 1419 | 99 | ||
GenBank mRNA | AK227229.1 | - | 2 - 3 | 1307 | 99 | |
BT002529.1 | - | 2 - 3 | 1306 | 99 | ||
BT010870.1 | - | 2 - 3 | 1220 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |