Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115252.3 | NP_190960.1 | 7 - 8 | 1391 | 84 | |
GenBank mRNA | AF015544.1 | - | 7 - 8 | 1240 | 84 | |
BT030362.1 | - | 7 - 8 | 1231 | 84 | ||
D89994.1 | - | 7 - 8 | 1360 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |