Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115271.5 | NP_190979.1 | 6 - 7 | 929 | 85 | |
GenBank mRNA | AF146226.1 | - | 6 - 7 | 709 | 85 | |
AY056272.1 | - | 6 - 7 | 890 | 85 | ||
AY091185.1 | - | 6 - 7 | 709 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |