Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATVPS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161207.1 | NP_001154679.1 | 12 - 13 | 1024 | 177 | |
NM_115343.4 | NP_567009.1 | 11 - 12 | 1089 | 129 | ||
GenBank mRNA | AF357527.1 | - | 11 - 12 | 909 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |