Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIL1-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125370.1 | NP_001118842.1 | 3 - 4 | 893 | 98 | |
NM_115353.5 | NP_191056.2 | 3 - 4 | 927 | 98 | ||
GenBank mRNA | AJ271376.1 | - | 3 - 4 | 951 | 98 | |
AK318844.1 | - | 3 - 4 | 895 | 98 | ||
AY093115.1 | - | 3 - 4 | 893 | 98 | ||
BT000172.1 | - | 3 - 4 | 875 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |