Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115402.4 | NP_191104.1 | 4 - 5 | 715 | 71 | |
GenBank mRNA | AF375426.1 | - | 4 - 5 | 418 | 71 | |
AY074822.1 | - | 4 - 5 | 324 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |