Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPGEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115424.3 | NP_191125.1 | 6 - 7 | 1512 | 129 | |
GenBank mRNA | BT020279.1 | - | 6 - 7 | 1395 | 129 | |
BT021117.1 | - | 6 - 7 | 1318 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |