Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339744.1 | NP_001325527.1 | 1 - 2 | 783 | 83 | |
NM_115432.5 | NP_191133.1 | 2 - 3 | 661 | 83 | ||
NM_202715.2 | NP_974444.1 | 2 - 3 | 645 | 83 | ||
GenBank mRNA | AY081292.1 | - | 2 - 3 | 482 | 83 | |
BT008750.1 | - | 2 - 3 | 308 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |