Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339748.1 | NP_001319763.1 | 13 - 14 | 2164 | 66 | |
NM_001339749.1 | NP_001326016.1 | 11 - 12 | 2431 | 66 | ||
NM_001339750.1 | NP_001326018.1 | 12 - 13 | 2353 | 66 | ||
NM_001339751.1 | NP_001326017.1 | 14 - 15 | 1642 | 66 | ||
NM_115443.4 | NP_567027.2 | 14 - 15 | 1648 | 66 | ||
NM_202716.3 | NP_974445.2 | 15 - 16 | 1861 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |