Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115457.3 | NP_191158.1 | 2 - 3 | 988 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |