Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203183.1 | NP_001190112.1 | 4 - 5 | 876 | 74 | |
NM_115553.4 | NP_191253.1 | 3 - 4 | 953 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |