Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDP1-LIKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115573.4 | NP_191273.1 | 3 - 4 | 1807 | 82 | |
NM_202720.2 | NP_974449.1 | 2 - 3 | 1859 | 82 | ||
GenBank mRNA | AK117921.1 | - | 3 - 4 | 1678 | 82 | |
BT005969.1 | - | 2 - 3 | 1567 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |