Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339858.1 | NP_001319779.1 | 20 - 21 | 2387 | 81 | |
NM_001339860.1 | NP_001325645.1 | 20 - 21 | 3152 | 81 | ||
NM_001339861.1 | NP_001325651.1 | 20 - 21 | 3206 | 81 | ||
NM_001339862.1 | NP_001325648.1 | 20 - 21 | 2821 | 81 | ||
NM_001339863.1 | NP_001325647.1 | 20 - 21 | 2859 | 81 | ||
NM_001339864.1 | NP_001325650.1 | 20 - 21 | 2367 | 81 | ||
NM_001339865.1 | NP_001325649.1 | 19 - 20 | 2523 | 81 | ||
NM_001339866.1 | NP_001325646.1 | 19 - 20 | 2561 | 81 | ||
NM_115601.3 | NP_567048.1 | 20 - 21 | 2425 | 81 | ||
GenBank mRNA | AK318626.1 | - | 19 - 20 | 2378 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |