Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115612.3 | NP_191311.1 | 13 - 14 | 2196 | 90 | |
NM_202722.1 | NP_974451.1 | 13 - 14 | 2121 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |