Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI-4KBETA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120971.3 | NP_196497.1 | 2 - 3 | 2291 | 140 | |
GenBank mRNA | AK228931.1 | - | 2 - 3 | 2277 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |