Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120977.3 | NP_568213.2 | 3 - 4 | 992 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |