Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC64-V | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120979.3 | NP_196504.2 | 7 - 8 | 1288 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |