Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121591.5 | NP_197090.2 | 6 - 7 | 1183 | 101 | |
NM_203057.1 | NP_974786.1 | 6 - 7 | 712 | 101 | ||
GenBank mRNA | AY045829.1 | - | 6 - 7 | 804 | 101 | |
AY091367.1 | - | 6 - 7 | 685 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |