Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203099.2 | NP_974828.1 | 1 - 2 | 309 | 76 | |
GenBank mRNA | AK220598.1 | - | 1 - 2 | 292 | 76 | |
AK221000.1 | - | 1 - 2 | 292 | 76 | ||
AY088224.1 | - | 1 - 2 | 292 | 76 | ||
BT025631.1 | - | 1 - 2 | 119 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |