Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344487.1 | NP_001331886.1 | 16 - 17 | 2242 | 63 | |
NM_001344488.1 | NP_001331887.1 | 16 - 17 | 2084 | 63 | ||
NM_123716.3 | NP_199163.1 | 17 - 18 | 2063 | 63 | ||
NM_203147.2 | NP_974876.1 | 17 - 18 | 2109 | 63 | ||
GenBank mRNA | AY057568.1 | - | 17 - 18 | 1878 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |