Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124376.4 | NP_568717.2 | 6 - 7 | 1130 | 65 | |
NM_203189.2 | NP_974918.1 | 6 - 7 | 998 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |