Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345181.1 | NP_001330467.1 | 9 - 10 | 1455 | 63 | |
NM_125000.4 | NP_568836.2 | 10 - 11 | 1334 | 63 | ||
NM_203211.1 | NP_974940.1 | 10 - 11 | 1397 | 63 | ||
GenBank mRNA | AF507916.1 | - | 10 - 11 | 1227 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |