Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345181.1 | NP_001330467.1 | 10 - 11 | 1575 | 87 | |
NM_125000.4 | NP_568836.2 | 11 - 12 | 1454 | 87 | ||
NM_203211.1 | NP_974940.1 | 11 - 12 | 1517 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |