Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345181.1 | NP_001330467.1 | 1 - 2 | 447 | 72 | |
NM_125000.4 | NP_568836.2 | 1 - 2 | 440 | 72 | ||
NM_203211.1 | NP_974940.1 | 1 - 2 | 363 | 72 | ||
GenBank mRNA | AF507916.1 | - | 1 - 2 | 333 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |