Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125247.4 | NP_200668.1 | 3 - 4 | 858 | 105 | |
NM_203232.2 | NP_974961.1 | 3 - 4 | 832 | 105 | ||
GenBank mRNA | AK117382.1 | - | 3 - 4 | 742 | 105 | |
BT005096.1 | - | 3 - 4 | 706 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |