Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125287.5 | NP_568894.1 | 7 - 8 | 1062 | 69 | |
NM_203233.1 | NP_974962.1 | 6 - 7 | 1530 | 69 | ||
GenBank mRNA | AB021706.1 | - | 7 - 8 | 803 | 69 | |
AY088200.1 | - | 7 - 8 | 959 | 69 | ||
JQ937241.1 | - | 7 - 8 | 788 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |