Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203683.1 | NP_001190612.1 | 2 - 3 | 1612 | 62 | |
NM_125835.5 | NP_201244.2 | 2 - 3 | 1614 | 62 | ||
NM_203261.2 | NP_974990.1 | 2 - 3 | 1614 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |