Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125859.4 | NP_568990.1 | 1 - 2 | 244 | 59 | |
NM_203262.3 | NP_974991.1 | 1 - 2 | 244 | 59 | ||
NM_203263.2 | NP_974992.1 | 1 - 2 | 149 | 59 | ||
GenBank mRNA | AF375435.1 | - | 1 - 2 | 180 | 59 | |
AY074827.1 | - | 1 - 2 | 111 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |