Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333075.1 | NP_001322563.1 | 3 - 4 | 732 | 74 | |
NM_103118.4 | NP_174658.1 | 3 - 4 | 638 | 74 | ||
GenBank mRNA | AK176629.1 | - | 3 - 4 | 638 | 74 | |
U40856.1 | - | 3 - 4 | 621 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |