Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MES14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103121.4 | NP_174661.1 | 3 - 4 | 993 | 93 | |
GenBank mRNA | AF424607.1 | - | 3 - 4 | 982 | 93 | |
AK226919.1 | - | 3 - 4 | 795 | 93 | ||
BT025240.1 | - | 3 - 4 | 641 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |