Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MES14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103121.4 | NP_174661.1 | 4 - 5 | 1124 | 128 | |
GenBank mRNA | AF424607.1 | - | 4 - 5 | 1113 | 128 | |
AK226919.1 | - | 4 - 5 | 926 | 128 | ||
BT025240.1 | - | 4 - 5 | 772 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |