Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123940.1 | NP_001117412.1 | 3 - 4 | 671 | 114 | |
NM_103198.3 | NP_564452.1 | 3 - 4 | 881 | 114 | ||
GenBank mRNA | AY059876.1 | - | 3 - 4 | 759 | 114 | |
AY093358.1 | - | 3 - 4 | 499 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |