Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333166.1 | NP_001322303.1 | 4 - 5 | 1725 | 69 | |
NM_103275.1 | NP_174811.1 | 4 - 5 | 1590 | 69 | ||
GenBank mRNA | DQ446324.1 | - | 4 - 5 | 1590 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |