Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC75-I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103285.1 | NP_174821.1 | 6 - 7 | 936 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |