Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103440.4 | NP_174980.3 | 12 - 13 | 1403 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |