Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ty2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103481.4 | NP_175021.2 | 2 - 3 | 311 | 153 | |
GenBank mRNA | AY128276.1 | - | 2 - 3 | 309 | 153 | |
BT014871.1 | - | 2 - 3 | 259 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |