Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103536.4 | NP_175076.2 | 3 - 4 | 879 | 119 | |
GenBank mRNA | BT002389.1 | - | 3 - 4 | 726 | 119 | |
BT009680.1 | - | 3 - 4 | 417 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |