Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333243.1 | NP_001320473.1 | 2 - 3 | 257 | 97 | |
NM_103587.3 | NP_175127.1 | 4 - 5 | 789 | 97 | ||
GenBank mRNA | AY069911.1 | - | 4 - 5 | 588 | 97 | |
AY093977.1 | - | 4 - 5 | 453 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |