Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333243.1 | NP_001320473.1 | 3 - 4 | 347 | 71 | |
NM_103587.3 | NP_175127.1 | 5 - 6 | 879 | 71 | ||
GenBank mRNA | AY069911.1 | - | 5 - 6 | 678 | 71 | |
AY093977.1 | - | 5 - 6 | 543 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |