Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATE2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103684.3 | NP_175222.1 | 1 - 2 | 473 | 84 | |
GenBank mRNA | AF242581.1 | - | 1 - 2 | 272 | 84 | |
AJ417834.1 | - | 1 - 2 | 246 | 84 | ||
AY045636.1 | - | 1 - 2 | 333 | 84 | ||
AY058231.1 | - | 1 - 2 | 233 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |