Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115736.2 | NP_191433.1 | 10 - 11 | 1339 | 116 | |
GenBank mRNA | AY099611.1 | - | 10 - 11 | 1339 | 116 | |
BT002169.1 | - | 10 - 11 | 1275 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |