Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115800.6 | NP_567084.1 | 4 - 5 | 589 | 74 | |
GenBank mRNA | BT033079.1 | - | 4 - 5 | 550 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |