Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035819.1 | NP_001030896.1 | 10 - 11 | 4176 | 73 | |
NM_001203209.1 | NP_001190138.1 | 11 - 12 | 4103 | 73 | ||
NM_001339998.1 | NP_001326481.1 | 11 - 12 | 4171 | 73 | ||
NM_115839.4 | NP_191536.2 | 11 - 12 | 4142 | 73 | ||
GenBank mRNA | AY227252.1 | - | 10 - 11 | 3936 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |